Would you like to log in again?
Nitric Oxide Synthase (NOS) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB211086-100TEST
75832-952EA 789.06 USD InStock
75832-952
Nitric Oxide Synthase (NOS) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simple, fast, fluorometric assay to screen/study/charecterize potential NOS inhibitors
  • Screening, studying, and characterizing potential NOS inhibitors
  • Generates a stable signal
  • Simple and high-throughput adaptable method
Nitric oxide synthases (EC 1.14.13.39) (NOS) is a family of enzymes that catalyze the production of nitric oxide (NO). In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. Nitric oxide plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. In contrast to its beneficial effects, NO has also been associated with numerous pathological situations such as hypotension accompanying septic shock, essential hypertension, and atherosclerosis. The overproduction of NO has been found to be the fundamental cause underlying neurodegenerative disorders and neuropathic pain. Therefore, developing small molecules for inhibition of NOS is therapeutically desirable. BioVision’s Nitric Oxide Synthase Inhibitor Screening Kit provides a simple and high-throughput adaptable method to screen/study/characterize potential NOS inhibitors. In this assay, nitric oxide generated by NOS undergoes a series of reactions and reacts with the fluorescent probe to generate a stable signal at Ex/Em = 360/450 nm, which is directly proportional to NOS activity. In the presence of a NOS-specific inhibitor, the formation of NO is reduced/abolished resulting in decrease or total loss of the fluorescence.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty