Would you like to log in again?
Total Bile Acids (TBA) Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB239702-100TEST
75832-954EA 1074.14 USD InStock
75832-954
Total Bile Acids (TBA) Assay Kit (Colorimetric), BioVision
Assays Immunological Assays
A simple, sensitive & easy colorimetric assay to quantitate total bile acids.
  • Estimation of Bile Acids in various biological samples
  • Can detect as little as 1 µM of Bile Acids in a variety of samples
  • Sensitive and specific
Bile Acids (BA) make 67% of the total composition of Bile. They are 24-carbon steroids generated during cholesterol metabolism. They form conjugates with either glycine or taurine to form bile salts. Five of the BAs account for more than 99% of the total population found in biofluids. The average composition in healthy individuals includes conjugates of cholic, chenodeoxycholic, deoxycholic and lithocholic acids. Bile acids are critical due to their ability to solubilize lipids by forming micelles with cholesterol, and fatty acids. Their synthesis is not only critical for the removal of cholesterol from the body abut they are also needed for proper uptake of dietary lipids into the small intestine. The measurement of circulatory Total Bile Acids (TBA) therefore provides information about hepatic functions and liver diseases such as jaundice, and hepatocellular injury. TBA estimation can detect liver damage during early stages and permits patients to get treatment before hepatic damages become irreversible. In addition, Bile Acids participate as signaling molecules interacting with G-protein coupled receptors (GPCR), TGR5, and nuclear receptor farnesoid X receptor (FXR). BioVision’s Total Bile Acid Assay Kit provides a simple, sensitive, and high-throughput adaptable approach to detect physiological concentration of total bile acids in a variety of biological fluids. The principle of the assay is based on an enzymatic cycling method in the presence of NADH and a chromophore. The reduction of the chromophore produces a stable colorimetric product the absorbance of which can be followed kinetically at 405 nm. This absorbance is directly proportional to the amount of TBA in the sample. Our assay is very specific and sensitive. Other metabolites found in biofluids do not interfere with the assay. The assay can detect as little as 1 µM of Bile Acids in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty