Would you like to log in again?
D-2-Hydroxyglutarate (D2HG) Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB211070-100TEST
75832-960EA 1116.92 USD InStock
75832-960
D-2-Hydroxyglutarate (D2HG) Assay Kit (Colorimetric), BioVision
Assays Cellular Assays
A fast, sensitive, easy to use and HTP adaptable colorimetric assay to measure D2HG in various biological samples.
  • Measurement of D2HG level in various cells, tissues, and biological fluids
  • Strong absorbance
  • Fast, sensitive, easy-to-use
In eukaryotic cells, Isocitrate Dehydrogenase (IDH1, IDH2 and IDH3) catalyzes the interconversion of Isocitrate and α-Ketoglutarate. In human cancers, an IDH mutation causes a gain-of-function, which reduces its affinity for isocitrate and facilitates the conversion of α-ketoglutarate to D-2-Hydroxyglutarate in the presence of NADP. D-2-Hydroxyglutarate (D2HG) is present at low level in normal cells and tissues, but is significantly elevated in metabolic diseases and various cancers. Therefore, detection of elevated D2HG is important for early diagnosis, prognosis and the development of therapeutic strategies against these maladies. BioVision’s D-2-Hydroxyglutarate Assay Kit provides a convenient method to detect D2HG in biological samples. In this assay, D-2-Hydroxyglutarate is oxidized to α-Ketoglutarate in the presence of D2HG Enzyme and Substrate Mix. The intermediate reduces the probe to a colored product with strong absorbance at 450 nm. This absorbance is proportional to the amount of D2HG present in the samples. This assay kit is fast, sensitive, easy to use and high-throughput adaptable. It can measure D-2-Hydroxyglutarate levels less than 10 µM in various samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty