Would you like to log in again?
6-Phosphogluconic Acid (6-PGA) Assay Kit (Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB211071-100TEST
75833-282EA 1116.27 USD InStock
75833-282
6-Phosphogluconic Acid (6-PGA) Assay Kit (Colorimetric), BioVision
Assays Cellular Assays
A simple, sensitive & easy colorimetric assay to detect 6-Phosphogluconate (6-PGA) levels lower than 20 µM.
  • Measurement of 6-Phosphogluconic Acid in various tissues/cells
  • Analysis of Pentose Phosphate Pathway and Entner-Doudoroff Pathway
  • Simple, sensitive, easy-to-use
6-Phosphogluconate (6-PGA) is an intermediate of both Pentose Phosphate Pathway (PPP) and Entner-Doudoroff Pathway. It is produced by the hydrolysis of 6-Phosphogluconolactone, catalyzed by 6-Phosphogluconolactonase. In the Pentose Phosphate Pathway, 6-PGA is utilized by 6-Phosphogluconate Dehydrogenase to generate ribulose-5-Phosphate and NADPH. These products are important for nucleic acid synthesis and various anabolic processes. In Prokaryotes, 6-Phosphogluconate is the main metabolite of Entner-Doudoroff pathway, and is converted into Pyruvate using both 6-Phosphogluconate Dehydratase and 2-Keto-3-Deoxyphosphogluconate aldolase. Recent studies show that long-term exposure to glucose perturbs the Pentose Phosphate Pathway, causes significant accumulation of 6-Phosphogluconate and impairs beta cell function. Measurement of 6-Phosphogluconate levels therefore is important for evaluating Pentose Phosphate Pathway, developing therapeutic approaches for diabetes research, and analyzing the Entner-Doudoroff Pathway in bacteria. BioVision’s 6-Phosphogluconate assay kit can be used with a variety of sample types. In this assay, 6-Phosphogluconate is converted to Ribulose-5-Phosphate by 6-Phosphogluconate Dehydrogenase in the presence of NAD, to form NADH, which reduces a probe and generates strong absorbance at 450 nm. This 6-Phosphogluconate Assay Kit is simple, sensitive & easy to use and can detect 6-Phosphogluconate levels lower than 20 µM.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty