Would you like to log in again?
Glucose-6-Phosphate Dehydrogenase Inhibitor Screening Kit (Colorimetric), BioVision Inc.
Supplier: BioVision
AB283371-100TEST
75833-288EA 949.95 USD InStock
75833-288
Glucose-6-Phosphate Dehydrogenase Inhibitor Screening Kit (Colorimetric), BioVision Inc.
Assays Protein Assays
Industry's first kit to screen, study, and characterize potential inhibitors of G6PDH.
  • Screening, characterizing, and studying potential inhibitors of Glucose-6-Phosphate Dehydrogenase
  • High-throughput adaptable and can be performed in less than 30 minutes
  • Sensitive, quick, and easy method for screening potential inhibitors
Glucose-6-Phosphate Dehydrogenase (G6PDH) is a cytosolic enzyme which plays a key role in the Pentose Phosphate Pathway (PPP). The PPP pathway supplies reducing energy to cells (such as erythrocytes) by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). The NADPH in turn maintains the level of glutathione in these cells that helps protect the red blood cells against oxidative damage. Recently studies have found increased G6PDH activity in various human tumors and knockdown of this G6PDH decreases cell proliferation and enhances apoptosis in vitro. Therefore, targeting this key enzyme is essential for developing novel therapeutic approach for treatment/cure of cancer. In BioVision’s G6PDH assay kit, Glucose-6-Phosphate is oxidized by glucose-6-phosphate dehydrogenase to generate 6-Phosphoglucono-δ-Lactone and form NADH, which reacts with a probe to produce a signal at OD 450 nm. In the presence of G6PDH inhibitor, these reactions are impeded, thus decreasing the rate and/or extent of generation of G6PDH-dependent absorbance. Thus, this kit provides a sensitive, quick, and easy method for screening potential inhibitors of G6PDH. G6PDH Inhibitor Control is included to compare the efficacy of test inhibitors. The assay is high-throughput adaptable and can be performed in less than 30 minutes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty