Would you like to log in again?
Tryptophan Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB211098-100TEST
75832-866EA 1032.65 USD InStock
75832-866
Tryptophan Assay Kit (Fluorometric), BioVision Inc.
Assays Protein Assays
Measurement of Tryptophan in biological fluids, including free and bound tryptophan in serum.
  • Simple, highly sensitive assay, HTS
  • Specific reactions
  • No interference from amino acids
Tryptophan (TRP) is one of the eight essential amino acids. In general, TRP is the least abundant amino acid in humans. Chemically, TRP side chain (indole) confers its unique fluorometric properties. External sources of tryptophan include chicken, tuna, bananas, cheese, chocolate etc. Tryptophan is mainly used for protein synthesis. Additionally, TRP serves as a building block for several metabolites including kynurenine, serotonin, tryptamine, melatonin, niacin, and NAD/NAPD. TRP is the only amino acid that can be found in blood in two forms: bound (BTRP) and free (FTRP) Tryptophan. Changes in tryptophan concentrations are directly related to a number of physiological and behavioral processes including: sleep, memory, depression, motion sickness, bipolar disorders, and schizophrenia. BioVision’s Tryptophan Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects tryptophan concentration in biological fluids, including free and bound tryptophan in serum. The principle is based on the non-enzymatic reaction to generate a fluorophore (Ex/Em = 370/440 nm) using tryptophan as a building block. The reaction is specific and other amino acids do not interfere with the assay. The assay can detect as little as 2.5 µM of TRP in a variety of biological samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty