Would you like to log in again?
Cysteine Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211099-100TEST
75832-868EA 799.84 USD InStock
75832-868
Cysteine Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Simple, Highly Sensitive, HTS adaptable assay to measure cysteine in a variety of biological samples.
  • Estimation of Cysteine in various biological samples
  • Can detect as little as 10 µM of Cysteine
  • Based on the cleavage of thiol group
Cysteine (CYS) is a sulfhydryl-containing amino acid and an important structural and functional part of proteins. In animals, cysteine is synthesized from transsulfuration of homocysteine, which is itself derived from metabolism of the amino acid methionine. Cystathionine β-synthase catalyzes condensation of homocysteine with serine to form cystathionine, which is deaminated and hydrolyzed by Cystathionine β-lyase to form cysteine and α-ketobutyrate. Because of its nucleophilic nature, the thiol group of cysteine has numerous biological functions. The formation of disulfide linkages between the thiol groups of cysteine residues helps to stabilize the tertiary and quaternary structure of proteins. Cysteine, homocysteine (HCY), and other aminothiols exist in plasma in reduced, oxidized, and protein-bound forms, interacting with each other through redox pathways. Cysteine is the limiting precursor of the major intracellular antioxidant glutathione. The individuals with lower cysteine levels are more prone to damage from reactive oxygen species, which are generally removed either by thiols or by glutathione-linked enzymes. An elevated level of total cysteine also predicts adverse outcomes such as cardiovascular diseases and metabolic syndromes. BioVision’s Cysteine Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects physiological concentration of cysteine in a variety of biological fluids. The principle of the assay is based on the cleavage of thiol group of reduced cysteine producing a fluorophore (Ex/Em = 365/450 nm) with a stable signal, which is directly proportional to the amount of total cysteine in the sample. The assay is specific and other thiol-based amino acids do not interfere with the assay. The assay can detect as little as 10 µM of Cysteine in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty