Would you like to log in again?
HIV-1 Protease Activity Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211105-100TEST
75832-882EA 896.32 USD InStock
75832-882
HIV-1 Protease Activity Assay Kit (Fluorometric), BioVision Inc.
Assays Immunological Assays
Sensitive fluorometric assay to detect HIV-1 Protease activity.
  • Sensitive fluorometric assay to detect HIV-1 Protease activity
  • Utilizes the ability of active HIV-1 protease
  • Can detect HIV-1 in samples and of purified enzyme
Human Immunodeficiency Virus (HIV) is the cause of the Acquired Immunodeficiency Syndrome (AIDS). HIV-1 protease is a retroviral aspartyl protease (retropepsin) that is essential for the life-cycle of the virus as it cleaves newly synthesized polyproteins at the appropriate places to create the mature protein components of an infectious HIV virion. Without effective HIV1 protease, HIV virions remain non-infectious. BioVision’s HIV-1 Activity assay kit utilizes the ability of active HIV-1 protease to cleave a synthetic peptide substrate to release the free fluorophore which can be easily quantified (Ex/Em = 330/450 nm) using a fluorometer or fluorescence microplate reader. This assay kit is simple, rapid and can detect HIV-1 protease activity in samples and of purified enzyme.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty