Would you like to log in again?
Hexokinase (HK) Inhibitor Screening Kit (Colorimetric), BioVision Inc.
Supplier: BioVision
AB211107-100TEST
75832-886EA 1049.55 USD InStock
75832-886
Hexokinase (HK) Inhibitor Screening Kit (Colorimetric), BioVision Inc.
Assays Enzyme Assays
Kit for screening Hexokinase Inhibitors.
A simple and high-throughput adaptable assay kit
Colorimetrically screen, study and characterize potential inhibitors of Hexokinase within 30 minutes
Simple, high-throughput adaptable
Hexokinases (HK) play an important role in glucose metabolism. Hexokinases phosphorylate glucose and generate glucose-6-phosphate for glycolysis. Hexokinases have four isoforms (HK-I, II, III and IV). HK-I, HK-II and HK-III have low Km, while HK-IV has 100 fold high Km. Hexokinase deficiency leads to severe human diseases such as X-linked muscular dystrophy and a rare autosomal recessive hemolytic anemia. On the other hand, increased hexokinase activity is detected in various tumors and is associated with metastasis. In BioVision’s Hexokinase Inhibitor Screening kit, glucose is converted to glucose-6-phosphate by hexokinase; the glucose-6-phosphate is oxidized by glucose-6-phosphate dehydrogenase to form NADH, which reduces a colorless probe to a colored product with strong absorbance at 450 nm. In the presence of a Hexokinase inhibitor, the reaction is impeded/abolished resulting in decrease or total loss of absorbance. This assay kit can be used to screen, study and characterize the potential inhibitors of Hexokinase. The assay is simple, high-throughput adaptable and can be performed within 30 min.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty