Would you like to log in again?
MMP-9 Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB284517-100TEST
75832-896EA 910.25 USD InStock
75832-896
MMP-9 Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
A Fluorometric Assay Kit to easily screen, study and charecterize MMP-9 inhibitors.
  • A Fluorometric Assay Kit (Ex/Em=325/393 nm) to screen/study/charecterize MMP-9 inhibitors easily. 100 assays
Matrix metalloproteinase (MMP) family proteins are involved in the breakdown of extracellular matrix, and have been recognized as potential targets for a variety of pathologies. Matrix metallopeptidase 9 (MMP-9), also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B, is the mostly studied MMP, due to its fundamental roles in cancer biology, autoimmune disease, and other conditions. MMP-9 is a metalloenzyme, composed of a pro-domain, a gelatin binding domain, a catalytic domain, a proline rich linker region, and a carboxyl terminal hemopexin like domain. This enzyme degrades various substrates including gelatin, collagen types IV / V, and elastin. BioVision’s MMP-9 Inhibitor Screening Kit provides a quick and sensitive way for screening, studying and characterizing potential inhibitors of MMP-9. In this assay, MMP-9 hydrolyzes a FRET-based MMP-9 substrate and releases the quenched fluorescent group Mca, which can be detected fluorometrically at Ex/Em=325/393 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty