Would you like to log in again?
EZScreen™ Lactate Colorimetric Assay Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB282923-384TEST
75832-906EA 827.23 USD InStock
75832-906
EZScreen™ Lactate Colorimetric Assay Kit, BioVision
Assays Cellular Assays
BioVision’s EZScreen™ Lactate Colorimetric Assay Kit uses an enzyme mix which reacts with L-Lactate specifically.
  • Kit detects 0.5-500 µM lactate samples
  • D-Lactate does not react with the enzyme mix
  • Formed product is directly proportional to the amount of L-Lactate present in the sample
Abnormal high concentration of lactate in blood has been related to many important diseases such as diabetes and lactate acidosis. Elevated Lactate concentration in blood is also associated with shock, post cardiac arrest, tissue ischemia, thiamine deficiency, liver disorder, mitochondrial diseases and many more. Therefore detection and quantitation of lactate in from blood is a diagnostic key for several diseases. BioVision’s EZScreen™ Lactate Colorimetric Assay Kit uses an enzyme mix which reacts with L-Lactate specifically. D-Lactate does not react with the enzyme mix. The enzymatic reaction with L-lactate produces a product that generates a color (OD= 590 nm) when reacts with the Lactate probe. The formed product is directly proportional to the amount of L-Lactate present in the sample. The method is quantitative, rapid, simple, sensitive, and designed for high throughput format. The kit provides a convenient means for detecting 0.5 to 5 mM Lactate in biological samples such as in blood, blood circulation, in cells, in culture media, in fermentation media etc. in 384 HTP format
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty