Would you like to log in again?
EZAlbumin™ Depletion Kit (Spin Column), BioVision
Supplier: BioVision
AB241023-25TEST
75832-914EA 543.74 USD InStock
75832-914
EZAlbumin™ Depletion Kit (Spin Column), BioVision
Protein Extraction Protein Extraction Kits
Convenient method to deplete or remove abundant albumin protein from samples
  • Removal of albumin from plasma, serum, antibody or other samples
  • Purification of Cibacron Blue 3G-A binding proteins
  • Purification of albumin
BioVision’s Albumin Depletion Kit provides easy and user-friendly spin columns for depletion/removal of abundant albumin protein from samples thus enhancing the sample analysis. The kit is based on the interaction of Cibacron Blue 3G-A (immobilized on beads) with albumin. Each spin column can bind ~2-3 mg of albumin and processes sample in less than one hour. After albumin depletion, samples can be used for subsequent ELISA, immunoassays and other downstream analyses such as gel electrophoresis and functional assays. These columns can also be used for purification and enrichment of a variety of Cibacron Blue dye-binding proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty