Would you like to log in again?
Alpha-Klotho (Soluble) (Human) ELISA, IBL-America
Supplier: IBL America
27998
102501-052EA 1119.63 USD InStock
102501-052
Alpha-Klotho (Soluble) (Human) ELISA, IBL-America
Assays ELISAs
Solid phase sandwich ELISA for the determination of human soluble α-Klotho in serum and EDTA-plasma.
  • For research use only, not for use in diagnostic procedures
  • α-Klotho is an extremely down-regulated gene
  • Important molecule within a living organism
  • Can measure Human soluble alpha-Klotho protein in human blood
Alpha-klotho” was identified as an extremely down-regulated gene in the genetically-modified mouse of which phenotype is very similar to various symptoms of human aging. Then, the sequences of Alpha-klotho genes have been identified in various species including humans based on the one of mouse. Alpha-Klotho protein is a 130 kDa, one-transmembrane protein and its expression is confirmed in the kidneys and the parathyroid glands. In recent years, it has become clear that alpha-Klotho is an important molecule within a living organism regulating the metabolism of mineral such as calcium and phosphorus. Therefore, it is considered that, in alpha-klotho mouse, early aging-like symptoms are induced by impaired mineral homeostasis caused by decreased expression of alpha-Klotho. Meanwhile, it is reported that the long N-terminal extracellular domain which comprises the major portion of sequence of Alpha-Klotho protein is released free into blood by shedding. However, there are many unclear points about functions and changes in concentration of free (soluble) alpha-Klotho protein, so it has been required to develop the measurement system of alpha-Klotho. This ELISA kit can measure Human soluble alpha-Klotho protein in human blood.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty