Would you like to log in again?
GLP-1 Active form (High sensitivity) ELISA, IBL-America
Supplier: IBL America
27700
103123-444EA 1419.29 USD InStock
103123-444
GLP-1 Active form (High sensitivity) ELISA, IBL-America
Assays ELISAs
Solid phase sandwich enzyme-linked immunosorbent assay (ELISA) for the determination of GLP-1 (Active Form) in human, mouse and rat EDTA-plasma, and cell culture supernatant.
  • For research use only, not for use in diagnostic procedures
  • Can quantitatively and specifically measure GLP-1
  • GLP-1 transcriptional product of proglucagon gene as same as glucagon
GLP-1 (Glucagon Like Peptide-1) is a transcriptional product of proglucagon gene as same as glucagon and one of gastrointestinal hormone which is secreted from L cells of lower section of the small intestine. GLP-1 (7-36) amide and GLP-1 (7-37) are active form and these peptides are dissolved in blood by DDP4 (dipeptidyl peptidase-4) into GLP-1 (9-36) amide and GLP-1 (9-37) and these peptides lose i ts activities in short period. GLP-1 is known as incretin as same as GIP (Glucose-dependent Insulinotropic Polypeptide), it promotes secretion of insulin from β cells of the isles of Langerhans in the pancreas as glucose dependently. GLP-1 also suppresses secretion of glucagon from α cell of pancreas and concentration of glucose in blood after meal. Drug of type 2 diabetes has been developed by utilizing the functional mechanism of GLP-1 such as blood sugar control through pancreas, delayed gastric emptying and appetite suppression which contributes weight loss as extra-pancreatic effects. This ELISA kit can quantitatively and specifically measure GLP-1, active form (GLP-1(7-36) amide and GLP-1 (7-37)).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty