Would you like to log in again?
Human GIP ELISA Kit, IBL-America
Supplier: IBL America
27203
102966-884EA 1419.29 USD InStock
102966-884
Human GIP ELISA Kit, IBL-America
Assays ELISAs
Solid, phase sandwich ELISA for the determination of human total GIP in EDTA-plasma and cell culture supernatant.
  • For research use only, not for use in diagnostic procedures
  • Detects both types of human GIP, active form (1-42) and inactivated form (3-42)
  • Can measure total GIP in samples
Incretins are a group of gastrointestinal hormones that cause an increase in the amount of insulin released from the beta cells of the islets of Langerhans after eating and they also inhibit glucagon release from the alpha cells of the Islets of Langerhans. GIP, typical incretin like GLP-1, was isolated and sequenced from intestinal mucosa as “gastric inhibitory peptide” in 1970, and then it was renamed as “glucose-dependent insulinotropic peptide”. It has been reported that GIP receptor is expressed in cells such as beta cell of pancreas, adipocyte or osteoblastic cell, and it plays essential roles in reserving of ingested nutrients within the body in each cell, and the control of GIP signal can lead to improvement of metabolic syndrome. GIP is rapidly inactivated to GIP (3-42) from active form of GIP (1-42) by DPP-IV in blood. This ELISA kit detects both types of human GIP, active form (1-42) and inactivated form (3-42), and can measure total GIP in samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty