Would you like to log in again?
Difco™ Actinomycete Isolation Agar, BD
Supplier: BD Diagnostics
212168 212168
90000-510EA 494.13 USD InStock
CA90000-510 90000-510
Difco™ Actinomycete Isolation Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Actinomycete Isolation Agar is used with added glycerol for isolating and cultivating actinomycetes from soil and water.
  • Sodium caseinate is a source of nitrogen
  • Sodium propionate is a substrate in anaerobic fermentation
  • Dipotassium phosphate provides buffering to maintain pH balance
  • Magnesium sulfate and ferrous sulfate provide sulfates and metallic ions
  • Agar is the solidifying agent
The added glycerol is a source of carbon to promote growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty