Would you like to log in again?
Difco™ Azide Blood Agar Base, BD
Supplier: BD Life Sciences
240920 240920
90003-164EA 459.89 USD InStock
90003-164 CA90003-164
Difco™ Azide Blood Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Azide Blood Agar Base is used for isolating streptococci and staphylococci and, supplemented with blood, for determining hemolytic reactions.
  • Peptones and beef extract provide essential nutrients
  • Sodium chloride maintains osmotic balance
  • Sodium azide is selective by inhibiting gram-negative bacteria
  • Agar is the solidifying agent
Supplementation with 5-10% blood provides additional growth factors for fastidious microorganisms, and is used to determine hemolytic patterns of bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty