Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylmercury hydroxide ~1M in aqueous solution

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Methylmercury(II) hydroxide

80-2300-10MMOL
100208-972EA 515.2 USD InStock
100208-972
Methylmercury hydroxide ~1M in aqueous solution
Methylmercury hydroxide


Formula: CH₄HgO
MW: 232.632 g/mol
MDL Number: MFCD00013924
CAS Number: 1184-57-2
UN: 2024
ADR: 6.1,II
Order Now
Specification Test Results
Chemical Formula CH3HgOH
Color and Form Colorless Liquid
Safety Hazardous

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty