Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-chloropyridine ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 5-Chloropyridin-2-amine5-Chloro-2-pyridylamine

2010025GM
100264-266EA 52.31 USD InStock
100264-266
2-Amino-5-chloropyridine ≥98%, crystalline powder
2-Amino-5-chloropyridine
  • White crystalline powder
  • Store in a cool, dry place


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 131…138 °C
MDL Number: MFCD00006324
CAS Number: 1072-98-6
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/96/21285596.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty