Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Asp(OtBu)-OH ≥99%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: D-Aspartic acid 4-tert-butyl ester

043591GM
100281-228EA 143.85 USD InStock
100281-228
H-D-Asp(OtBu)-OH ≥99%, crystals
H-D-Asp(OtBu)-OH
  • White crystals
  • Store at 0-8 °C


Formula: C₈H₁₅NO₄
Melting Pt: >195 °C
MDL Number: MFCD00077101
CAS Number: 64960-75-4
ADR: ,
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/00/21286400.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty