Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sweet Almonds Beta-Glucosidase
Supplier: MP Biomedicals
0219519705 0219519725 0219519780
IC0219519725EA 143.69 USD InStock
IC0219519725 IC0219519705 IC0219519780
Sweet Almonds Beta-Glucosidase
Enzymes
β-Glucosidase is a glucosidase enzyme
  • Used in synthesis of glucosides and fucosides
  • Potential applications in pharmaceuticals, cosmetic and detergent industries
  • One unit is the amount of enzyme which liberates 1.0 μg of glucose per minute at 35°C, pH 5.0
GHS Symbol(s): GHS08
Signal word: Danger
a-Amylase: <0.000001%
It is an exocellulase with specificity for a variety of β-D-glycoside substrates. It is involved in the hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides. They convert cellobiose and cellooligosaccharides produced by the endo and exoglucanases to glucose.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty