Would you like to log in again?
Surgipath® Bulk Processing Cassettes, Leica
Supplier: Leica
Surgipath®
3802256 3802258 3802240 3802250 3802252 3802244 3802246 3802242 3802245 3802257 3802248 3802247 3802254
75810-238CS 212.7 USD InStock
75810-238 75810-246 75810-236 75809-270 75809-260 75809-262 75809-264 75809-266 75810-244 75810-234 75810-242 75809-268 75810-240
Surgipath® Bulk Processing Cassettes, Leica
Embedding Cassettes Tissue Processing Cassettes
Processing Cassettes are precision molded using solvent-resistant plastic.
  • Available in 13 colors
  • 50° writing surface
  • Compatible with most metal lids and base molds
Features include a three-sided textured writing surface and easy flow-through vents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty