Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid solution 1%, Surgipath®

Supplier: Leica Biosystems
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

Surgipath®
38016SS1B
75810-386EA 103.18 USD InStock
75810-386
Acetic acid solution 1%, Surgipath®
Acetic acid
Component of the Leica Gomorri's Green or Blue Collagen Trichrome Stain Kits

  • Used as a differentiator in Trichrome Stains


Formula: H₃CCOOH
MW: 60.05 g/mol
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty