Would you like to log in again?
Surgipath® Bouins Solution, Leica
Supplier: Leica
Surgipath®
38016SS1A
75810-384EA 158.57 USD InStock
75810-384
Surgipath® Bouins Solution, Leica
Stains Microscopy and staining reagents
Component of the Leica Gomorri's Green or Blue Collagen Trichrome Stain Kits.
  • Acts as mordant
  • Attaches dye to tissue
Acts as a mordant to strongly attach subsequent dye molecules to the tissue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty