Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tetra-Chloroauric(III) acid solution

Supplier: Leica Biosystems
Synonyms: Gold chlorideAuric chlorideGold trichloridetetra-Chloroauric acidHydrogen tetrachloroaurate (III)

38016SS12E
75810-378EA 662.29 USD InStock
75810-378
tetra-Chloroauric(III) acid solution
tetra-Chloroauric (III) acid
Solution Gold chloride, component of the Leica Modified Grocott's Methenamine Silver Stain Kit.

  • Used as a toning solution


Formula: HAuCl₄
MW: 339.78 g/mol
MDL Number: MFCD00011322
CAS Number: 16903-35-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty