Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromic acid modified

Supplier: Leica Biosystems
Danger

Surgipath®
38016SS12A
75810-370EA 249.7 USD InStock
75810-370
Chromic acid modified
Chromic acid
Modified Chromic Acid Solution component of the Leica Modified GMS Silver Kit.

Oxidizes polysaccarides of the fungal walls into aldehydes.


Formula: CrH₂O₄
MW: 118.01 g/mol
CAS Number: 7738-94-5
UN: 1755
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty