Would you like to log in again?
Surgipath® Modified GMS Silver Stain Kit , Leica
Supplier: Leica
Surgipath®
38016SS12
75810-368KT 980.48 USD InStock
75810-368
Surgipath® Modified GMS Silver Stain Kit , Leica
Stains Histology Stains
The Modified GMS Silver Stain Kit is intended for use in histological observation of fungi, basement membrane and some opportunistic organisms such as pneumocystis carinii in tissue specimens.
Ordering information: This kit contains 500 mL each of Modified Chromic Acid, Sodium Bisulfite 1% Aqueous Solution, Light Green Stain, Gold Chloride, and Sodium Thiosulfate 2% Aqueous Soltion. Also included is 250 mL of Silver Nitrate 0.5% Aqueous Solution. Due to refrigeration, Methanemine Borax Solution must be ordered separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty