Would you like to log in again?
Surgipath® Snowcoat® Micro Slides for Cytology, Leica
Supplier: Leica
Surgipath®
3808381
75809-518CS 131.51 USD InStock
75809-518
Surgipath® Snowcoat® Micro Slides for Cytology, Leica
Slides Microscope Slides
Snowcoat® Micro Slides with two circles are for use with cytology funnels.
  • For cytology
  • Two circles match cytology funnel
  • For non-gyn specimens
These slides are 3”x 1” White Snowcoat® slides with two circles, each having a diameter of 1/2". These circles match up with the Cytology Funnel, which is to be used for preparation of non-gyn cytology specimens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty