Would you like to log in again?
Difco™ PALCAM Antimicrobic Supplement, BD Life Sciences
Supplier: BD Life Sciences
263710 263710
90003-744EA 326.21 USD InStock
CA90003-744 90003-744
Difco™ PALCAM Antimicrobic Supplement, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
PALCAM Antimicrobic Supplement is added to PALCAM Medium Base for isolating and cultivating Listeria, particularly from foods and milk products.
  • Presence of ceftazidime increases selectivity of the medium
  • Lyophilized appearance: White, free-flowing, homogeneous powder
  • Rehydrated appearance: Colorless solution
Ceftazidime found in PALCAM Antimicrobic Supplement helps effectively suppress growth of most commonly occurring non-Listeria species of bacteria present in foods. The ceftazidime concentration is reduced from 20 mg/L to 8 mg/L for improved growth and recovery of Listeria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty