Would you like to log in again?
Sealing Tape, Associated Bag
Supplier: Associated Bag
110-45C
10030-704CT 359.34 USD InStock
10030-704
Sealing Tape, Associated Bag
Tapes
With resilient polypropylene backing and hot-melt adhesive, this sealing tape is designed for use with industrial tape machines.
  • Tough Polypropylene Backing
  • Hot-Melt Adhesive
  • Clear
  • Excellent Performance in High-Volume Shipping Rooms
  • Machine-Length
It performs well in a variety of environmental conditions and in high-volume shipping rooms.
Packaging: Six per Carton. Weight: 26 lb.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty