Would you like to log in again?
Quick-Load Purple 50 bp DNA Ladder, New England Biolabs
Supplier: New England Biolabs
N0556S N0556S
103218-930EA 99 USD InStock
103218-930 CA103218-930
Quick-Load Purple 50 bp DNA Ladder, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
Quick-load Purple 50 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye.
  • Convenient, ready-to-load format with NEB's proprietary purple loading dye
  • Size range: 50 to 1350 bp
  • Sharp bands
  • No UV Shadow
  • Easy to identify reference bands
Tracking dye does not leave a shadow under UV light. The Quick-Load Purple 50 bp DNA Ladder consists of a proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 17 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The 200 and 500 bp bands have increased intensity to serve as reference points.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty