Would you like to log in again?
Exonuclease VIII
Supplier: New England Biolabs
M0545S M0545S
103218-960EA 82 USD InStock
103218-960 CA103218-960
Exonuclease VIII
Enzymes
Exonuclease VIII, truncated, is a genetically engineered active domain of exonuclease VIII from E.coli. Exonuclease VIII, truncated is able to initiate nucleotide removal from the 5' termini of linear double-stranded DNA in the 5' to 3' direction (1). The enzyme does not degrade supercoiled dsDNA and circular ssDNA.
  • Double-stranded DNA specific exonuclease
  • Initiates at the 5' termini of linear double-stranded DNA
  • Catalyzes the removal of nucleotides from linear double-stranded DNA in the 5' to 3' direction
  • Removal of linear double-stranded DNA, leaving behind circular DNA in the sample
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty