Would you like to log in again?
Alkaline Peptone Water, Hardy Diagnostics
Supplier: Hardy Diagnostics
K03
10063-764PK 149.66 USD InStock
10063-764
Alkaline Peptone Water, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
An enrichment medium for the cultivation of Vibrio spp
An enrichment medium for the cultivation of Vibrio spp. from feces and other materials.
from feces and other materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty