Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blood Agar, 5%, Slant, Hardy Diagnostics

Supplier: Hardy Diagnostics
L12
10153-942PK 97.33 USD InStock
10153-942
Blood Agar, 5%, Slant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Blood Agar, 5%, Slant, 5.5mL, 16x100mm tube, order by the package of 20

Tryptic Soy Agar with 5% Sheep Blood For the cultivation of micro-organisms.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty