Would you like to log in again?
HardyDisks™ AST Cefotaxime, CTX-30, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z8221
10790-416EA 34.66 USD InStock
10790-416
HardyDisks™ AST Cefotaxime, CTX-30, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Cefotaxime, CTX-30, HardyDisks™ AST, for susceptibility testing
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty