Would you like to log in again?
FID (Fluorescence-Indole-Dentrification) Agar Slant , Hardy Diagnostics
Supplier: Hardy Diagnostics
Y21
75803-036PK 206.29 USD InStock
75803-036
FID (Fluorescence-Indole-Dentrification) Agar Slant , Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the identification of non-fermentative bacilli by fluorescein pigment production, nitrate reduction, and indole production.
For the identification of non-fermentative bacilli by fluorescein pigment production, nitrate reduction, and indole production.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty