Would you like to log in again?
KF Streptococcus Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G376
75802-990PK 44.6 USD InStock
75802-990
KF Streptococcus Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Recommended for the selective isolation and enumeration of fecal streptococci
Recommended for the selective isolation and enumeration of fecal streptococci (including Enterococcus) from water and food samples by direct culture or membrane filtration.
This product is not intended to be used for the diagnosis of human disease.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty