Would you like to log in again?
Martin Lewis with Lincomycin, Hardy Diagnostics
Supplier: Hardy Diagnostics
E31 E31
10789-390PK 57.65 USD InStock
10789-390 CA10789-390
Martin Lewis with Lincomycin, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A selective medium for the recovery of Neisseria gonorrhoeae from both genital and oropharyngeal specimens.
Hardy Diagnostics Martin Lewis with Lincomycin is a selective medium for the recovery of Neisseria gonorrhoeae from both genital and oropharyngeal specimens. It is used in qualitative procedures for the isolation of Neisseria gonorrhoeae with suppression of most other gram-negative diplococci, gram-negative bacilli, gram-positive organisms, and yeast. The CO2 enriched environment allows for the growth of pathogenic Neisseria spp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty