Would you like to log in again?
Microgen® Bioproducts Bacillus ID Panel, Hardy Diagnostics
Supplier: Hardy Diagnostics
MID66
10032-748EA 789.13 USD InStock
10032-748
Microgen® Bioproducts Bacillus ID Panel, Hardy Diagnostics
Microbiology Test Systems Inhibitor Test Systems
For the mesophilic Bacillus species and related genera associated with food poisoning and food spoilage.
A 24 substarate microwell identification system for the mesophilic Bacillus species and related genera associated with food poisoning and food spoilage
Microgen Bacillus-ID identification system consists of two microwell strips (labeled BAC 1 and BAC 2), each containing 12 dehydrated substrates for the performance of either carbohydrate fermentation tests or other biochemical based tests. The last well in the second strip is a carbohydrate fermentation control well for use as a reference well in the interpretation of these tests. The selection of the substrates included in the test panel has been determined using computer based analysis of all available substrates for the identification or differentiation of this group of organisms.
Identification of isolates is achieved by recording the results visualized by a color change after 24 and 48 hours of incubation at 30°C and the addition of appropriate reagents (Indole, Nitrate and VP tests) after 48 hours.
These results are then analyzed using the Microgen Identification System Software. This software is available for free to Microgen users upon request. Reagents required, but not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty