Would you like to log in again?
Moeller's Decarboxylase Base, Hardy Diagnostics
Supplier: Hardy Diagnostics
Y41
10693-094PK 188.05 USD InStock
10693-094
Moeller's Decarboxylase Base, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of gram negative enteric bacilli.
For the differentiation of gram negative enteric bacilli based on the decarboxylation reaction of amino acids. A control tube for use with Y42, Y43, and Y44.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty