Would you like to log in again?
Moeller's Lysine Decarboxylase, Hardy Diagnostics
Supplier: Hardy Diagnostics
Y43
10693-096PK 240.47 USD InStock
10693-096
Moeller's Lysine Decarboxylase, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of gram negative enteric bacilli.
For the differentiation of gram negative enteric bacilli based on the decarboxylation reaction of amino acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty