Would you like to log in again?
Phenol Red Broth with Dextrose, Hardy Diagnostics
Supplier: Hardy Diagnostics
Y304 Y304
10124-528PK 210.56 USD InStock
CA10124-528 10124-528
Phenol Red Broth with Dextrose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of microorganisms by carbohydrate fermentation.
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty