Would you like to log in again?
Potato Dextrose Agar with Chloramphenicol, Hardy Diagnostics
Supplier: Hardy Diagnostics
W94BX W94BX
10789-406PK 288 USD InStock
10789-406 CA10789-406
Potato Dextrose Agar with Chloramphenicol, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Recommended for the selective cultivation of fungi from mixed samples.
Recommended for the selective cultivation of fungi from mixed samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty