Would you like to log in again?
Potato Dextrose Agar with Chlortetracycline, Hardy Diagnostics
Supplier: Hardy Diagnostics
W93 W93
89501-564PK 49.4 USD InStock
CA89501-564 89501-564
Potato Dextrose Agar with Chlortetracycline, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Recommended for use in the cultivation of yeast and mold.
Recommended for use in the cultivation of yeast and mold and for the inducement of sporulation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty