Would you like to log in again?
Rose Bengal Agar with Chloramphenicol 1.5X, Hardy Diagnostics
Supplier: Hardy Diagnostics
Q81
10158-952PK 439.57 USD InStock
10158-952
Rose Bengal Agar with Chloramphenicol 1.5X, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of fungi from environmental and food sources.
For the selective isolation and enumeration of fungi from environmental and food sources.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty