Would you like to log in again?
XLT4 Supplement, Hardy Diagnostics
Supplier: Hardy Diagnostics
235310
75803-060EA 129.36 USD InStock
75803-060
XLT4 Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Dehydrated culture media supplement
Is a dehydrated culture media supplement. Use this product with XLT-4 Agar Base.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty