Would you like to log in again?
Virtua™ V4 Protective Eyewear, 3M™
Supplier: 3M
Virtua™
7000029998 7000029999 11671-00000-20
89095-232CS 112.61 USD InStock
89095-232 89095-230 89095-234
Virtua™ V4 Protective Eyewear, 3M™
Safety Glasses
Virtua™ V4 protective eyewear combines versatility and value for a comfortable, contoured fit.
  • Lightweight and unisex
  • Soft, dual-injected temples relieve hot spots around the ears
  • Wraparound design shields eyes for ful protection
  • Contoured temples fit comfortably and snugly without pressure points
  • Impact-resistant polycarbonate lens absorbs 99.9% UV rays
This economy line of safety glasses combines style, protection and affordability to safeguard workers on jobsites. These safety glasses are recommended for use in chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding, and sawing applications.
Industries in which these safety glasses are commonly used include aviation, chemicals, construction, food and beverage, general manufacturing, mining, oil and gas, pharmaceuticals, primary metals, and transportation.
Certifications: ANSI Z87.1-2003.
Caution: Do not use this eyewear in an industrial setting without contacting a supervisor or safety specialist to insure they provide proper protection for the specific workplace hazards. Some operations may require special filter lenses, machine guards, goggles, face shields, or other safety equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty