Would you like to log in again?
Clamp Holders, Chemglass
Supplier: Chemglass
CG-2040-C-01
30622-652EA 64.15 USD InStock
30622-652
Clamp Holders, Chemglass
Clamps General Purpose Clamps
Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance
Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjusment and secure clamping power. Useful wherever clamping at 90° is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty