Would you like to log in again?
Vernier Emissions Fiber
Supplier: VERNIER SOFTWARE & TECH SE
VSP-EM-FIBER
470116-642EA 93.99 USD InStock
470116-642
Vernier Emissions Fiber
Cables
Fiber optic cable for Vernier spectometer.
  • Glass core material
  • 2.2 mm PVC covering
  • SMA 905 connectors
  • 400 µm core diameter
This fiber is an accessory for the Vernier emissions spectrometer that allows for precise measurements of emissions spectrum wavelengths. Fiber can also be used with the Vernier spectrometer and other Ocean Optics VIS-NIR spectrometers.
Not compatible with Vernier SpectroVis Plus. To do emissions with an SpectroVis Plus Spectrophotometer, use SpectroVis Optical Fiber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty