Would you like to log in again?
EVA® Women's Pink Rimless Safety Glasses, PIP
Supplier: Protective Industrial Products
250-10-0920
10120-243CS 2059.19 USD InStock
10120-243
EVA® Women's Pink Rimless Safety Glasses, PIP
Safety Glasses
The first safety eyewear designed specifically for women in the workforce.
  • Smaller and narrower than standard safety glasses; allows the eyewear to better fit the facial structure of a woman
  • Pink rubber co-molded temples and molded nose bridge provide a more secure fit
  • Lightweight, rimless design provides all day user comfort
  • 10 Base curve lens
  • A portion of the proceeds are donated to the American Cancer Society
Clear Lens.
Anti-Scratch Coating
Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching.
Anti-Fog Coating
Eliminates the condensation of moisture on lenses that causes fogging, ensuring that lenses and vision stay clear.
Certifications: Meets ANSI Z87.1+ standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty